Thermo Scientific Chemicals

Ethidium Bromide, Molecular Biology Grade, 98%

Catalog number: J67270.14
25 g, Each
Thermo Scientific Chemicals

Ethidium Bromide, Molecular Biology Grade, 98%

Catalog number: J67270.14
25 g, Each
Quantity
Catalog number: J67270.14
also known as J67270-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1239-45-8
IUPAC Name
3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
Molecular Formula
C21H20BrN3
InChI Key
ZMMJGEGLRURXTF-UHFFFAOYSA-N
SMILES
[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Form
Powder or crystals or crystalline powder
Infrared spectrum
Conforms
pH
4.0-7.0
Assay from Suppliers CofA
≥97.5%
Appearance (Color)
Red to purple

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text