Thermo Scientific Chemicals

Chloramphenicol, 10 mg/ml in ethanol, sterile-filtered

Catalog number: J67273.AB
10 mL, Each
Thermo Scientific Chemicals

Chloramphenicol, 10 mg/ml in ethanol, sterile-filtered

Catalog number: J67273.AB
10 mL, Each
Quantity
Catalog number: J67273.AB
also known as J67273-AB
Price (USD)
Have Questions?

Chemical Identifiers

CAS
56-75-7
IUPAC Name
2,2-dichloro-N-[1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
Molecular Formula
C11H12Cl2N2O5
InChI Key
WIIZWVCIJKGZOK-UHFFFAOYNA-N
SMILES
OCC(NC(=O)C(Cl)Cl)C(O)C1=CC=C(C=C1)[N+]([O-])=O
Appearance (Color)
Colorless
Comment
Product Description states 10 mg/mL composition
Insoluble material
None
Form
Liquid
Comment
Sterile

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text