6-Bromoindirubin-3'-oxime, 97%, Thermo Scientific Chemicals
6-Bromoindirubin-3'-oxime, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Bromoindirubin-3'-oxime, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
50 mg
Catalog number J67275.MA
also known as J67275-MA
Price (USD)/ Each
138.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
163.00 
Save 24.35 (15%)
-
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)/ Each
138.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
163.00 
Save 24.35 (15%)
Add to cart
6-Bromoindirubin-3'-oxime, 97%, Thermo Scientific Chemicals
Catalog numberJ67275.MA
Price (USD)/ Each
138.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
163.00 
Save 24.35 (15%)
-
Add to cart
Chemical Identifiers
CAS667463-62-9
IUPAC Name6'-bromo-3-(hydroxyamino)-1',2'-dihydro-[2,3'-biindolyliden]-2'-one
Molecular FormulaC16H10BrN3O2
InChI KeyWNWSUJQVZJJGLF-UHFFFAOYSA-N
SMILESONC1=C2C=CC=CC2=NC1=C1C(=O)NC2=C1C=CC(Br)=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink to red to dark red
Assay from Suppliers CofA≥96.0%
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only