Thermo Scientific Chemicals

Gold nanoparticles, 30nm, supplied in 0.1 mg/ml sodium citrate with stabilizer, OD1, 526nm absorption, Thermo Scientific Chemicals

Catalog number: J67284.AC
25 mL, Each
Thermo Scientific Chemicals

Gold nanoparticles, 30nm, supplied in 0.1 mg/ml sodium citrate with stabilizer, OD1, 526nm absorption, Thermo Scientific Chemicals

Catalog number: J67284.AC
25 mL, Each
Quantity
Catalog number: J67284.AC
also known as J67284-AC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-57-5
IUPAC Name
gold
Molecular Formula
Au
InChI Key
PCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES
[Au]
Form
Liquid
Assay (special)
Particles / ml: 1.7E ± 11
Assay (special)
Core size: 30nm ± 2
lambda-max (UV-Vis)
526nm

Description

For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text