Marimastat, 98%
Thermo Scientific Chemicals

Marimastat, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
1 mg
Catalog number J67288.MA
also known as J67288-MA
Price (USD)/ Each
295.00
-
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)/ Each
295.00
Add to cart
Marimastat, 98%
Catalog numberJ67288.MA
Price (USD)/ Each
295.00
-
Add to cart
Chemical Identifiers
CAS154039-60-8
IUPAC Name(2S,3R)-N'-[(1S)-2,2-dimethyl-1-(methylcarbamoyl)propyl]-N,2-dihydroxy-3-(2-methylpropyl)butanediamide
Molecular FormulaC15H29N3O5
InChI KeyOCSMOTCMPXTDND-OUAUKWLOSA-N
SMILESCNC(=O)[C@@H](NC(=O)[C@H](CC(C)C)[C@H](O)C(=O)NO)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Mass Spectrum: Consistant with structure
Assay (HPLC)≥ 98%
A broad spectrum inhibitor of matrix metalloproteases (MMPs); inhibits MMP-9, MMP-1, MMP-2, MMP-14 and MMP-7 with IC50 values of 3, 5, 6, 9 and 13 nM respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C