LY 294002, 98%
Thermo Scientific Chemicals

LY 294002, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
25 mg
Catalog number J67289.LB0
also known as J67289-#0
Price (USD)/ Each
92.90
-
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)/ Each
92.90
Add to cart
LY 294002, 98%
Catalog numberJ67289.LB0
Price (USD)/ Each
92.90
-
Add to cart
Specifications
Quantity5 mg
DescriptionWhite to pale yellow
Recommended StorageStore at -20°C
Assay98%
CAS154447-36-6
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C