Kanamycin, 50 mg/ml in distilled water, sterile-filtered
Kanamycin, 50 mg/ml in distilled water, sterile-filtered
Kanamycin, 50 mg/ml in distilled water, sterile-filtered
Kanamycin, 50 mg/ml in distilled water, sterile-filtered
Thermo Scientific Chemicals

Kanamycin, 50 mg/ml in distilled water, sterile-filtered

Kanamycin monosulfate, CAS # 25389-94-0, is an aminoglycoside sulfate salt with antibacterial activity. | CAS: 25389-94-0 | C18H38N4O15S | 582.575 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J67354.AD
also known as J67354-AD
Price (USD)
237.65
Special offer
Online exclusive
Ends: 26-Sep-2025
293.00
Save 55.35 (19%)
Each
In stock
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)
237.65
Special offer
Online exclusive
Ends: 26-Sep-2025
293.00
Save 55.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS25389-94-0
IUPAC Name(2R,3S,4S,5R,6R)-2-(aminomethyl)-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-3-{[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}oxane-3,4,5-triol; sulfuric acid
Molecular FormulaC18H38N4O15S
InChI KeyOOYGSFOGFJDDHP-KMCOLRRFSA-N
SMILESOS(O)(=O)=O.NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Comment50mg/ml aqueous solution is prepared in 18 megohm-cm ñ 1 water. The solution is filter sterilized using 0.22 micron filter
It has been used in tissue culture media. It is used as an additive in culture media for the isolation of group D streptococci on Kanamycin Esculin Azide Agar and for selection of transformed plant cells containing the neomycin phosphotransferase on a kanamycin-medium. It can also be used as a selection agent for cells transformed with kanamycin B resistance gene. It is recommended for use in cell culture applications at 100mg/mL. Kanamycin can be used as a selection agent in several types of isolation media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Kanamycin monosulfate is the sulfate salt form of kanamycin A
• It was isolated from Streptomyces kanamyceticus
• This compound inhibits translocation during protein biosynthesis. It binds to the 30S ribosomal subunit, causing misreading of the mRNA sequence

Application

• Kanamycin monosulfate shows antibacterial activity. It is effective against gram-positive and gram-negative bacteria, as well as Mycoplasma species
• This compound is also involved in translating prokaryotic ribosomes
• In molecular biology experiments, it is commonly used to select and isolate transformants expressing the kanamycin resistance gene, and plants that have the NPT II (APH3) gene incorporated

RUO – Research Use Only

General References:

  1. Patel, K. N.; Limgavkar, R. S.; Raval, H. G.; Patel, K. G.; Gandhi, T. R. High-Performance Liquid Chromatographic Determination of Cefalexin Monohydrate and Kanamycin Monosulfate with Precolumn Derivatization. J. Liq. Chromatogr. Related Technol. 2015, 38 (6), 716-721.
  2. Chi, Z.; Rong, Y. J.; Li, Y.; Tang, M. J.; Chi, Z. M. Biosurfactins production by Bacillus amyloliquefaciens R3 and their antibacterial activity against multi-drug resistant pathogenic E. coli. Bioprocess. Biosyst. Eng. 2015, 38 (5), 853-861.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link