Thermo Scientific Chemicals

DNA Molecular Weight Marker, 100bp Ladder, (100-1517 bp)

Catalog number: J67383.4EQ
1.25 mL, Each
Thermo Scientific Chemicals

DNA Molecular Weight Marker, 100bp Ladder, (100-1517 bp)

Catalog number: J67383.4EQ
1.25 mL, Each
Quantity
Catalog number: J67383.4EQ
Price (USD)
Have Questions?
Form
Liquid
Assay (special)
DNA Concentration (A260): 50-55 µg/mL (Conforms)
Assay (special)
Electrophoretic Pattern (Marker): Coforms
Assay (special)
Non-Specific DNase Activity (DNA, 16 hour): Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text