Thermo Scientific Chemicals

n-Octyl-beta-D-glucopyranoside, 98+%, Thermo Scientific Chemicals

Catalog number: J67390.06
5 g, Each
Thermo Scientific Chemicals

n-Octyl-beta-D-glucopyranoside, 98+%, Thermo Scientific Chemicals

Catalog number: J67390.06
5 g, Each
Quantity
Catalog number: J67390.06
also known as J67390-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29836-26-8
IUPAC Name
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol
Molecular Formula
C14H28O6
InChI Key
HEGSGKPQLMEBJL-RKQHYHRCSA-N
SMILES
CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
Appearance (Color)
White to pale cream
Assay (unspecified)
Related Substances (n-Octanol): ≤0.5%
Assay (special)
Alpha Isomer: ≤0.5%
Loss on Drying
≤0.5%
Assay from Supplier's CofA
≥98.0% (HPLC)

Description

It is used for the solubilization and reconstitution of membrane-bound proteins. The high critical micelle concentration (0.7%) of n-octylglucoside facilitates ready removal from final protein extracts by dialysis or gel filtration. n-Octylglucoside can be used in 2D electrophoresis and to improve selectivity of immunoprecipitation of phosphotyrosine modified proteins. Also useful in 2D electrophoresis and in an immunoprecipitation buffer for the immunopurification of phospho-tyrosine peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for the solubilization and reconstitution of membrane-bound proteins. The high critical micelle concentration (0.7%) of n-octylglucoside facilitates ready removal from final protein extracts by dialysis or gel filtration. n-Octylglucoside can be used in 2D electrophoresis and to improve selectivity of immunoprecipitation of phosphotyrosine modified proteins. Also useful in 2D electrophoresis and in an immunoprecipitation buffer for the immunopurification of phospho-tyrosine peptides.

Solubility
Soluble in DMSO, methanol and water.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Store under inert gas. Store in refrigerator at -20°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text