Thermo Scientific Chemicals
Gold nanoparticles, 40nm, supplied in 0.1mM PBS, 95%, OD1, 530 nm absorption, Thermo Scientific Chemicals
Catalog number: J67409.AC
25 mL, Each
Thermo Scientific Chemicals
Gold nanoparticles, 40nm, supplied in 0.1mM PBS, 95%, OD1, 530 nm absorption, Thermo Scientific Chemicals
Catalog number: J67409.AC
25 mL, Each
Quantity
Catalog number: J67409.AC
also known as J67409-AC
Price (USD)
Quantity
-
Have Questions?
Description
For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM