Gold nanoparticles, 5nm, biotin-functionalized, PEGylated, OD50, supplied in purified water, 515-520 nm absorption, Thermo Scientific Chemicals
Gold nanoparticles, 5nm, biotin-functionalized, PEGylated, OD50, supplied in purified water, 515-520 nm absorption, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold nanoparticles, 5nm, biotin-functionalized, PEGylated, OD50, supplied in purified water, 515-520 nm absorption, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mL
1 mL
Catalog number J67413.AMI
also known as J67413-&I
Price (USD)/ Each
335.00
-
Add to cart
Quantity:
0.5 mL
Request bulk or custom format
Price (USD)/ Each
335.00
Add to cart
Gold nanoparticles, 5nm, biotin-functionalized, PEGylated, OD50, supplied in purified water, 515-520 nm absorption, Thermo Scientific Chemicals
Catalog numberJ67413.AMI
Price (USD)/ Each
335.00
-
Add to cart
Specifications
Chemical Name or MaterialGold nanoparticles
ColorBrown to Red
Name Notebiotin to functionalized, PEGylated, OD: 50, supplied in purified water, 515 to 520nm absorption
CAS7440-57-5
Storage Note 12° to 8°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only