Boric acid, Molecular Biology Grade, 99.5% min
Boric acid, Molecular Biology Grade, 99.5% min
Thermo Scientific Chemicals

Boric acid, Molecular Biology Grade, 99.5% min

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J67419.36
also known as J67419-36
Price (USD)/ Each
66.90
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
66.90
Add to cart
Boric acid, Molecular Biology Grade, 99.5% min
Catalog numberJ67419.36
Price (USD)/ Each
66.90
-
Add to cart
Chemical Identifiers
CAS10043-35-3
IUPAC Nameboric acid
Molecular FormulaBH3O3
InChI KeyKGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILESOB(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy Metals(as Pb): ≤0.001%
Assay (unspecified)≥99.5%
DNaseNone detected
Insoluble material≤0.005%
Chloride content≤0.001%
View more
Used to promote amidations of carboxylic acids and amines in catalytic amounts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only