Citrate Buffer, 0.2M buffer soln., pH 4.0, low endotoxin
Citrate Buffer, 0.2M buffer soln., pH 4.0, low endotoxin
Thermo Scientific Chemicals

Citrate Buffer, 0.2M buffer soln., pH 4.0, low endotoxin

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
100 mL
Catalog number J67435.AK
also known as J67435-AK
Price (USD)/ Each
105.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
124.00 
Save 18.35 (15%)
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
105.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
124.00 
Save 18.35 (15%)
Add to cart
Citrate Buffer, 0.2M buffer soln., pH 4.0, low endotoxin
Catalog numberJ67435.AK
Price (USD)/ Each
105.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
124.00 
Save 18.35 (15%)
-
Add to cart
Chemical Identifiers
CAS6132-04-3
Specifications
Chemical Name or MaterialCitrate Buffer
Name Notelow endotoxin
CAS6132-04-3
ColorColorless
Storage Note 12° to 8°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only