Gold nanoparticles, 60nm, supplied in 0.1mM PBS, 95%, OD1, 540nm absorption, Thermo Scientific Chemicals
Gold nanoparticles, 60nm, supplied in 0.1mM PBS, 95%, OD1, 540nm absorption, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold nanoparticles, 60nm, supplied in 0.1mM PBS, 95%, OD1, 540nm absorption, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 mL
Catalog number J67438.AC
also known as J67438-AC
Price (USD)/ Each
88.60
-
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)/ Each
88.60
Add to cart
Gold nanoparticles, 60nm, supplied in 0.1mM PBS, 95%, OD1, 540nm absorption, Thermo Scientific Chemicals
Catalog numberJ67438.AC
Price (USD)/ Each
88.60
-
Add to cart
Specifications
Chemical Name or MaterialGold nanoparticles
ColorPink to Red
Name Notesupplied in 0.1mM PBS, 95%, OD: 1, 540nm absorption
CAS7440-57-5
Storage Note 12° to 8°C
View more
For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
RUO – Research Use Only