Gold nanoparticles, 250nm, supplied in 0.1mM PBS, reactant-free, 99%, Thermo Scientific Chemicals
Gold nanoparticles, 250nm, supplied in 0.1mM PBS, reactant-free, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold nanoparticles, 250nm, supplied in 0.1mM PBS, reactant-free, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 mL
Catalog number J67457.AC
also known as J67457-AC
Price (USD)/ Each
143.00
-
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)/ Each
143.00
Add to cart
Gold nanoparticles, 250nm, supplied in 0.1mM PBS, reactant-free, 99%, Thermo Scientific Chemicals
Catalog numberJ67457.AC
Price (USD)/ Each
143.00
-
Add to cart
Specifications
Chemical Name or MaterialGold nanoparticles
ColorBrown to Pink
Name Notesupplied in 0.1mM PBS, reactant-free
CAS7440-57-5
Storage Note 12° to 8°C
View more
Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only