Thermo Scientific Chemicals

Anisomycin, 10 mg/ml in DMSO, sterile-filtered

Catalog number: J67498.XF
1 mL, Each
Thermo Scientific Chemicals

Anisomycin, 10 mg/ml in DMSO, sterile-filtered

Catalog number: J67498.XF
1 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J67498.XF
also known as J67498-XF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22862-76-6
IUPAC Name
(2S,3R,4R)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl acetate
Molecular Formula
C14H19NO4
InChI Key
YKJYKKNCCRKFSL-BFHYXJOUSA-N
SMILES
COC1=CC=C(C[C@@H]2NC[C@@H](O)[C@@H]2OC(C)=O)C=C1
Form
Liquid
Composition
Anisomycin: 10 mg/mL in DMSO, sterile filtered

Description

It is applied as an antiparasitic agent and antineoplastic agent. It acts as a protein synthesis inhibitor (blocks translation), potent activator of stress-activated protein kinases (JNK/SAPK) and p38 MAP kinase. It also scts as a potent signaling agonist to selectively elicit homologous desensitization of immediate early gene induction (c-fos, fosB, c-jun, junB and junD). Activates mitogen-activated protein (MAP) kinases (JNK/SAPK and p38/RK).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an antiparasitic agent and antineoplastic agent. It acts as a protein synthesis inhibitor (blocks translation), potent activator of stress-activated protein kinases (JNK/SAPK) and p38 MAP kinase. It also scts as a p
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text