Tris-HCl, 0.5M buffer soln., pH 7.4, low endotoxin
Tris-HCl, 0.5M buffer soln., pH 7.4, low endotoxin
Thermo Scientific Chemicals

Tris-HCl, 0.5M buffer soln., pH 7.4, low endotoxin

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
100 mL
Catalog number J67501.AK
also known as J67501-AK
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
Add to cart
Tris-HCl, 0.5M buffer soln., pH 7.4, low endotoxin
Catalog numberJ67501.AK
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialTRIS-HCl
Name Notelow endotoxin
CAS1185-53-1
ColorColorless
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only