D-(+)-Cellopentaose, 95%, Thermo Scientific Chemicals
D-(+)-Cellopentaose, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-(+)-Cellopentaose, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
2 mg
5 mg
Catalog number J67561.MCR
also known as J67561-M^
Price (USD)/ Each
112.00
-
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (USD)/ Each
112.00
Add to cart
D-(+)-Cellopentaose, 95%, Thermo Scientific Chemicals
Catalog numberJ67561.MCR
Price (USD)/ Each
112.00
-
Add to cart
Chemical Identifiers
CAS2240-27-9
IUPAC Name(2R,3S,4R,5R)-5,6-dihydroxy-2,3,4-tris({[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy})hexanal
Molecular FormulaC24H42O21
InChI KeyMUIAGSYGABVSAA-XHFFWFORSA-N
SMILESOC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H](O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@@H](O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Purity≥94.0%
FormPowder or crystals or crystalline powder
Appearance (Color)White
Used by cellulosic bacteria as an energy source. Used to identify, differentiate and characterize oligosaccharide metabolizing enzymes such as endoglucanases and --glucosidase(s). Can also be used in saccharification and ethanol fermentation research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used by cellulosic bacteria as an energy source
  • Used to identify, differentiate and characterize oligosaccharide metabolizing enzymes such as endoglucanases and _x001A_-glucosidase(s)
  • Can also be used in saccharification and ethano
RUO – Research Use Only