Thermo Scientific Chemicals

Ringer's Solution, lactate-buffered

Catalog number: J67572.AP
500 mL, Each
Thermo Scientific Chemicals

Ringer's Solution, lactate-buffered

Catalog number: J67572.AP
500 mL, Each
Quantity
Catalog number: J67572.AP
also known as J67572-AP
Price (USD)
Quantity
-
Have Questions?
Form
Liquid
Comment
All components are dissolved in UV treated 18.2 megohm-cm ± 1 water, followed by pH adjustment and filtered through 0.22 micron filter.
Composition
100mM sodium chloride, 3.7mM potassium chloride, 1.3mM calcium chloride, 1.2mM magnesium sulfate, 28mM sodium bicarbonate, and 20mM sodium lactate
Appearance (Color)
Clear colorless
pH
7.4 ± 0.15

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text