Thermo Scientific Chemicals

D-(+)-Cellotetraose, 95%

Catalog number: J67583.MA
10 mg, Each
Thermo Scientific Chemicals

D-(+)-Cellotetraose, 95%

Catalog number: J67583.MA
10 mg, Each
Quantity
Catalog number: J67583.MA
also known as J67583-MA
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38819-01-1
IUPAC Name
(2S,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6S)-6-{[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Formula
C24H42O21
InChI Key
LUEWUZLMQUOBSB-YQGOCCRESA-N
SMILES
OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@H]4[C@H](O)[C@@H](O)C(O)O[C@@H]4CO)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
Form
Powder or crystals or crystalline powder
Appearance (Color)
White to off-white
Purity
≥94.0%

Description

Derived from cellulose degradation. Used by cellulosic bacteria as an energy source. Useful for characterization and identification of oligosaccharide metabolizing enzymes such as endoglucanases and --glucosidases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Derived from cellulose degradation
  • Used by cellulosic bacteria as an energy source
  • Useful for characterization and identification of oligosaccharide metabolizing enzymes such as endoglucanases and _x001A_-glucosidases
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text