Thermo Scientific Chemicals

Sodium n-dodecyl sulfate, 99%, Molecular Biology Grade

Catalog number: J67606.22
100 g, Each
Thermo Scientific Chemicals

Sodium n-dodecyl sulfate, 99%, Molecular Biology Grade

Catalog number: J67606.22
100 g, Each
Quantity
Catalog number: J67606.22
also known as J67606-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
151-21-3
IUPAC Name
sodium dodecyl sulfate
Molecular Formula
C12H25NaO4S
InChI Key
DBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
pH
6.0-7.5 (1% solution)
UV/Vis
Absorbance 280 nm (3% in water): ≤0.4
Chloride content
≤0.10%
Solution Test
Passes (10% in water)
Assay from Suppliers CofA
≥99.0% (unspecified)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text