Thermo Scientific Chemicals

alpha-Cyano-4-hydroxycinnamic acid, Ultrapure MALDI Matrix, Thermo Scientific Chemicals

Catalog number: J67635.EXK
5 x 10 mg, Each
Thermo Scientific Chemicals

alpha-Cyano-4-hydroxycinnamic acid, Ultrapure MALDI Matrix, Thermo Scientific Chemicals

Catalog number: J67635.EXK
5 x 10 mg, Each
Quantity
Catalog number: J67635.EXK
also known as J67635-!K
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
28166-41-8
IUPAC Name
(2E)-2-cyano-3-(4-hydroxyphenyl)prop-2-enoic acid
Molecular Formula
C10H7NO3
InChI Key
AFVLVVWMAFSXCK-VMPITWQZSA-N
SMILES
OC(=O)C(=C\C1=CC=C(O)C=C1)\C#N
Form
Homogeneous crystals
GC/MS Analysis
Homogeneous crystals that lead to matrix for matrix-assisted laser desorption ionization (MALDI) mass spectra with excellent resolution, high sensitivity, and high S/N ratios.

Description

For use as a matrix in MALDI mass spectrometry analysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text