Thermo Scientific Chemicals
D-(+)-Cellotriose, 95%
Catalog number: J67702.MB
25 mg, Each
Thermo Scientific Chemicals
D-(+)-Cellotriose, 95%
Catalog number: J67702.MB
25 mg, Each
Quantity
Catalog number: J67702.MB
also known as J67702-MB
Price (USD)
Quantity
-
Have Questions?
Description
A 3-glucose cellulodextrin derived from cellulose degradation. An energy source for cellulosic bacteria. Can be used to help identify, differentiate and characterize oligosaccharide metabolizing enzymes such as endoglucanases and --glucosidases. Also useful in saccharification and ethanol fermentation research.