Thermo Scientific Chemicals
Hanks' Balanced Salt Solution (10X), with calcium, with magnesium, without phenol red
Catalog number: J67771.AP
500mL, Each
Thermo Scientific Chemicals
Hanks' Balanced Salt Solution (10X), with calcium, with magnesium, without phenol red
Catalog number: J67771.AP
500mL, Each
Quantity
Catalog number: J67771.AP
also known as J67771-AP
Price (USD)
Quantity
-
Have Questions?
Appearance (Color)
Clear colorless
Comment
Product comprises of two components: a salt solution and a metal solution. Metal solution containing calcium chloride and magnesium chloride proved as a separate solution.
Form
Liquid