Phosphate-buffered saline (DPBS, 1X), Dulbecco's formula, without calcium, without magnesium
Phosphate-buffered saline (DPBS, 1X), Dulbecco's formula, without calcium, without magnesium
Thermo Scientific Chemicals
Phosphate-buffered saline (DPBS, 1X), Dulbecco's formula, without calcium, without magnesium
Have Questions?
Change viewbuttonViewtableView
Capacity:
1 L
500 mL
Catalog number J67802.K2
also known as J67802-K2
Price (USD)/ Each
42.65
Online exclusive
47.40 
Save 4.75 (10%)
-
Add to cart
Capacity:
1 L
Request bulk or custom format
Price (USD)/ Each
42.65
Online exclusive
47.40 
Save 4.75 (10%)
Add to cart
Phosphate-buffered saline (DPBS, 1X), Dulbecco's formula, without calcium, without magnesium
Catalog numberJ67802.K2
Price (USD)/ Each
42.65
Online exclusive
47.40 
Save 4.75 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Capacity1 L
Packaging TypePlastic bottle
FormLiquid
Unit SizeEach
An anticoagulant and blood preservative. Permits the storage of whole blood at refrigerator temperatures for approximately 10 weeks.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Ambient temperatures