You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Water, RNase-Free, DEPC Treated, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Element Search
Structure Search
Thermo Scientific Chemicals
Water, RNase-Free, DEPC Treated, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Have Questions?
Change view
Quantity:
10 x 1 mL
25 mL
100 mL
500 mL
1 L
Packaging:
Nalgene Bottle
Try the chem dex
TM
web app
Catalog number
J70783.XCR
Price (USD)
/ Each
36.65
Online exclusive
41.10
Save 4.45 (11%)
-
Add to cart
Quantity:
10 x 1 mL
Packaging:
Nalgene Bottle
Request bulk or custom format
Price (USD)
/ Each
36.65
Online exclusive
41.10
Save 4.45 (11%)
Add to cart
Water, RNase-Free, DEPC Treated, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Catalog number
J70783.XCR
Price (USD)
/ Each
36.65
Online exclusive
41.10
Save 4.45 (11%)
-
Add to cart
Chemical Identifiers
CAS
7732-18-5
IUPAC Name
water
Molecular Formula
H2O
InChI Key
XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES
O
View more
Specifications
Specification Sheet
DNase
(endo): Passes test
RNase
Passes test
Comment
Contaminant Testing: Enzyme Inhibition: Passes test
DNase
(exo): Passes test
RUO – Research Use Only