Thermo Scientific Chemicals

Guanidine hydrochloride, 99.5+%, Molecular Biology Grade, Thermo Scientific Chemicals

Catalog number: J75823.22
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Guanidine hydrochloride, 99.5+%, Molecular Biology Grade, Thermo Scientific Chemicals

Catalog number: J75823.22
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: J75823.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-01-1
IUPAC Name
hydrogen guanidine chloride
Molecular Formula
CH6ClN3
InChI Key
PJJJBBJSCAKJQF-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC(N)=N
Assay (unspecified)
Absorbance (225 nm, 6M, 1 cm): ≤ 0.2
Assay (unspecified)
Ammeline: None detected
Identification (FTIR)
Conforms
Protease activity
Passes test
Trace Metal
Arsenic (As): ≤ 1 ppm

Description

  • Used for extraction of RNA.
  • Preparation of high molecular weight genomic DNA.
  • Solubilizing inclusion bodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used for extraction of RNA
  • Preparation of high molecular weight genomic DNA
  • Solubilizing inclusion bodies
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text