Urea, ≥99.5%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Urea, ≥99.5%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Urea, ≥99.5%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

Chaotropic agent used in denaturing polyacrylamide gel electrophoresis. When 8M urea is not sufficiently denaturing, 40% formamide may be included.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number J75826.A1
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
Add to cart
Urea, ≥99.5%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Catalog numberJ75826.A1
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS57-13-6
IUPAC Nameurea
Molecular FormulaCH4N2O
InChI KeyXSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILESNC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
DNasePasses test
FormWhite crystalline powder
Assay (unspecified)≥99.5%
Assay (unspecified)Cyanate: None detected
Chloride content≤ 0.0005%
View more
  • Chaotropic agent used in denaturing polyacrylamide gel electrophoresis.
  • When 8M urea is not sufficiently denaturing, 40% formamide may be included.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only