Thermo Scientific Chemicals

Ready-Made LB (Luria-Bertani) Broth Powder, Thermo Scientific Chemicals

Catalog number: J75852.30
250 g, Each
Thermo Scientific Chemicals

Ready-Made LB (Luria-Bertani) Broth Powder, Thermo Scientific Chemicals

Catalog number: J75852.30
250 g, Each
Quantity
Catalog number: J75852.30
Price (USD)
Have Questions?
Form
Free flowing powder
Composition
50% Casein peptone, 25% Yeast extract and 25% Sodium chloride
pH
(2.0%, H2O): 7.0 ? 0.2

Description

For the propagation of bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Off-white free flowing powder
  • pH(2.0%, H2O): 7.0 ± 0.2
Contents
  • Casein Peptone: 50%
  • Yeast Extract: 25%
  • Sodium Chloride: 25%
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text