Thermo Scientific Chemicals
TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Catalog number: J75892.K2
1 L, Nalgene bottle, Each
Thermo Scientific Chemicals
TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Catalog number: J75892.K2
1 L, Nalgene bottle, Each
Quantity
Packaging
Catalog number: J75892.K2
Price (USD)
Quantity
-
Have Questions?
pH
(25?C, 20X): 7.4 ? 0.1
DNase
(exo): Passes test
Protease activity
Passes test
RNase
Passes test
DNase
(endo): Passes test
Description
Used in Western blotting protocols.