Thermo Scientific™

Maxima Probe/ROX qPCR Master Mix (2X)

Catalog number: K0233
Have Questions?
Thermo Scientific™

Maxima Probe/ROX qPCR Master Mix (2X)

Catalog number: K0233
Have Questions?
Catalog Number
K0233
Unit Size
4,000 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
200 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1,000 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K02334,000 reactions
-
K0231200 reactions
-
K02321,000 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
**********************
Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of Maxima Probe qPCR Master Mix and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific Maxima Probe qPCR Master Mixes are ready-to-use solutions optimized for quantitative real-time PCR. The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added.

Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG) (see Reference 1). The use of Maxima Probe qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates.

Maxima Probe qPCR Master Mixes are compatible with most real-time thermal cyclers.

Highlights

• Specificity—Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
• Sensitivity—detects low copy number targets
• Wide linear range—accurate quantification across 9 orders of magnitude
• Reproducibility and convenience—ready-to-use 2X master mix

Applications

• Gene expression
• siRNA validation
• Genotyping
• Pathogen detection

Includes
Maxima Probe/ROX qPCR Master Mix (2X) includes Maxima Hot Start Taq DNA Polymerase and dNTPs (also dUTP) in an optimized PCR buffer supplemented with ROX passive reference dye. Supplied with nuclease-free water.

Related Products
Maxima Probe/ROX qPCR Master Mix (2X)
Maxima Probe qPCR Master Mix (2X), with separate ROX vial
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
Format
Tube
GC-Rich PCR Performance
High
PCR Method
qPCR
Polymerase
Taq DNA Polymerase
Reaction Speed
Standard
For Use With (Equipment)
7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
Product Line
Maxima
Product Type
qPCR Master Mix
Sufficient For
4000 Reactions of 25 μL
For Use With (Application)
Gene Expression
Concentration
2X
No. of Reactions
4,000 Reactions
Sample Type
DNA, dsDNA
Volume
4 x 12.5 mL

Contents & Storage

Contains:
• 4 x 12.5 mL Maxima Probe/ROX qPCR Master Mix (2X) containing Maxima Hot Start Taq DNA Polymerase and dNTPs (also dUTP) in an optimized PCR buffer with ROX passive reference dye; sufficient for 4000 x 25 μL reactions.
• 2 x 30 mL Nuclease-Free Water

Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text