Luminaris Color HiGreen qPCR Master Mix, low ROX
Product Image
Thermo Scientific™

Luminaris Color HiGreen qPCR Master Mix, low ROX

**********************Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
K0371250 Reactions
K03745000 Reactions
K03731250 Reactions
K0372500 Reactions
Catalog number K0371
Price (USD)/ Each
293.00
-
Add to cart
No. of Reactions:
250 Reactions
Price (USD)/ Each
293.00
Add to cart
Luminaris Color HiGreen qPCR Master Mix, low ROX
Catalog numberK0371
Price (USD)/ Each
293.00
-
Add to cart
**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of Luminaris Color HiGreen qPCR Master Mix and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific Luminaris Color HiGreen and Luminaris HiGreen qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR. The master mixes include Thermo Scientific Hot Start Taq DNA polymerase, uracil-DNA glycosylase (UDG) and dNTPs in an optimized PCR buffer. Only the template and primers need to be added. The SYBR Green I intercalating dye allows for DNA detection and analysis without using sequence-specific probes.

The Hot Start Taq DNA polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP and UDG are included in the mix for carry-over contamination control (see Reference 1). The Luminaris Color HiGreen qPCR Master Mixes are supplemented with an inert blue dye and a separate Yellow Sample Buffer that contains a yellow dye. The qPCR reaction mix containing both components is green.

The use of Luminaris Color HiGreen and Luminaris HiGreen qPCR Master Mixes in qPCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral and cDNA templates. Individual formulations of Luminaris Color HiGreen and Luminaris HiGreen qPCR Master Mixes with ROX at different concentrations or fluorescein as passive reference dyes are available for most qPCR platforms (see Real-time PCR instrument compatibility chart).

Highlights

• Blue master mix and Yellow Sample Buffer for easy pipetting
• UDG in the master mix to prevent carry-over contamination
• Equal performance of blue and colorless master mixes
• Versions with premixed ROX or fluorescein for different real-time platforms
• Standard cycling protocols
• Excellent qPCR performance:
Specificity: Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
Sensitivity: single copy detection
Wide linear range: accurate quantification across 9 orders of magnitude
Reproducibility and convenience: ready-to-use 2X master mix


Applications

• Gene expression analysis
• siRNA validation
• Genotyping
• Pathogen detection

Includes

Luminaris Color HiGreen Low ROX qPCR Master Mix (2X) includes Hot Start Taq DNA Polymerase, UDG and dNTPs (also dUTP) in an optimized PCR buffer with a blue dye. The master mix contains low concentration of ROX passive reference dye for specific real-time platforms (see Real-Time PCR instrument compatibility chart) and is supplied with 40X Yellow Sample Buffer and nuclease-free water.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodSYBR
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
PolymeraseTaq DNA Polymerase
Reaction SpeedStandard
For Use With (Equipment)7500 System, QuantStudio™, ViiA™ 7 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P
Product LineLuminaris
Product TypeqPCR Master Mix
Quantity250 reactions
Sufficient For250 Reactions
For Use With (Application)Gene Expression
Concentration2X
Fidelity (vs. Taq)2 X
No. of Reactions250 Reactions
Sample TypeDNA, dsDNA
Unit SizeEach
Contents & Storage
Contains:
• 2 x 1.25 mL Luminaris Color HiGreen Low ROX qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer with a blue dye; sufficient for 250 x 20 μL reactions. The master mix contains low concentration of ROX passive reference dye for specific real-time platforms.
• 1 x 1.25 mL 40X Yellow Sample Buffer
• 2 x 1.25 mL Nuclease-Free Water

Store at -20°C.