GeneJET Plasmid Maxiprep Kit
For better long-term performance store purification columns at 2°C to 8°C.
GeneJET Plasmid Maxiprep Kit
Thermo Scientific™

GeneJET Plasmid Maxiprep Kit

Thermo Scientific GeneJET Plasmid Maxiprep Kit, sufficient for 25 preps, utilizes silica-based membrane technology in the form of convenient spin column.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
K049110 Preps
K049225 Preps
Catalog number K0491
Price (EUR)
183,00
Each
-
Add to cart
No. of Reactions:
10 Preps
Price (EUR)
183,00
Each
Add to cart
Ask our AI about this Product

Thermo Scientific GeneJET Plasmid Maxiprep Kit, sufficient for 25 preps, utilizes silica-based membrane technology in the form of convenient spin column. Each prep recovers up to 750 μg of high copy plasmid DNA that can be used in a wide variety of molecular biology procedures, such as restriction endonuclease digestion, PCR, cloning, transformation, automated sequencing, in vitro transcription, and transfection of robust cell lines.

Highlights

  • Efficient—high yields up to 750 μg of high quality plasmid DNA from 250 mL of bacterial culture
  • High concentration of DNA purified— >400 ng/μL
  • Convenient—spin column format, centrifuge or vacuum protocols
  • Fast—procedure takes 60 minutes, considerably faster than using gravity flow columns

Applications

Isolated plasmid DNA is suitable for all conventional molecular biology procedures, including:

  • Transfection of robust cell lines
  • FastDigest restriction enzyme digestion
  • Automated fluorescent sequencing
  • PCR
  • Cloning
  • Transformation
  • In vitro transcription
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeSpin Column
Concentration400 ng/μL
For Use With (Application)Real-Time Quantitative PCR (qPCR), Transfection, Cloning, Sequencing, In Vitro Transcription, Transformation, Nucleic Acid Labeling, PCR
High-throughput CompatibilityNot High-throughput Compatible (Manual)
No. of Reactions10 Preps
Prep Scale> 200 μg (Large-Scale) Plasmid DNA
Product LineGeneJET
Product TypePlasmid MaxiPrep Kit
Quantity10 preps
Sample TypeBacteria
ScaleMaxi
TargetPlasmid DNA
Test Time60 min.
FormatSpin Column
Isolation TechnologySilica Column
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link