Search Thermo Fisher Scientific
货号 | 数量 |
---|---|
K0503 | 250 次反应 |
K0502 | 50 次制备 |
We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.
Yes, MagJET and GeneJET Plasmid DNA Purification Kits are suitable for plasmid DNA purification from EndA+ strains. Please refer to the specific kit protocol for details. GeneJET Plasmid Miniprep Kit requires Wash Solution I (Cat. No. R1611, not included in the kit) for EndA+ strains.
The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.
We recommend performing centrifugation steps at room temperature when the temperature is not specified.
No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.
分享货号、产品名称或链接