Thermo Scientific™

DecaLabel DNA Labeling Kit

Catalog number: K0622
Have Questions?
Thermo Scientific™

DecaLabel DNA Labeling Kit

Catalog number: K0622
Have Questions?
Catalog Number
K0622
Unit Size
30 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K062230 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific DecaLabel DNA Labeling Kit is an advanced system for fast synthesis of radiolabeled DNA probes of high specific activity. It's based on the improved random-primed method developed by Feinberg and Vogelstein.

The primary improvement over traditional random-primer kits involves the use of random decamers instead of hexamers to ensure more efficient annealing with DNA at 37°C. In addition, Klenow Fragment, exo- has been genetically engineered to remove exonuclease activity. The improved enzyme does not degrade the labeled probe during reaction and allows high labeling yields even from low template amounts. DNA fragments of any length can be uniformly labeled. Two labeling mixes are provided for flexibility in using either radiolabeled dATP or dCTP.

Highlights

High specific radioactivity of probes—greater than 1 x 109 dpm/μg DNA
Fast—5 minutes at 37°C
High yields with Klenow Fragment, exo no degradation of a labeled probe during reaction
Flexible—suitable for either radiolabeled dATP or dCTP

Applications

• Generation of radiolabeled DNA probes for use in a variety of hybridization experiments: Southern and Northern blots, colony/plaque hybridizations, dot/slot blots, and in situ hybridizations.

Includes

Klenow Fragment, exo-
• Decanucleotides in Reaction Buffer
• Mix A (-dATP)
• Mix C (-dCTP)
dNTP Mix
• Control Template
Water, nuclease-free
• Detailed Protocol

Principle
Random decamers are annealed to a denatured template DNA, and radiolabeled dNTPs are then incorporated into new DNA strands by Klenow Fragment, exo- (see Figure 1 under Supporting Data).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Radioactive
Format
Kit
Final Product Type
DNA
Labeling Target
Genomic DNA, PCR Products, DNA (General)
Includes Label or Dye
No
Labeling Method
Direct Labeling
No. of Reactions
30
Product Line
DecaLabel
Product Type
DNA Labeling Kit
Quantity
30 Reactions

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text