Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Product Type | Label or Dye |
---|---|---|
K06T04L025 | Conjugation Kit | NovaFluor™ Blue 725 |
K06T04L008 | Conjugation Kit | NovaFluor™ Blue 510 |
K06T04L002 | Conjugation Kit | NovaFluor™ Blue 530 |
K06T04L009 | Conjugation Kit | NovaFluor™ Blue 555 |
K06T04L010 | Conjugation Kit | NovaFluor™ Blue 585 |
K06T04L003 | Conjugation Kit | NovaFluor™ Blue 610/30S |
K06T04L011 | Conjugation Kit | NovaFluor™ Blue 610/70S |
K06T04L004 | Conjugation Kit | NovaFluor™ Blue 660/40S |
K06T04L021 | Conjugation Kit | NovaFluor™ Blue 690 |
K06T04L012 | Conjugation Kit | NovaFluor™ Blue 660/120S |
K06T04L026 | Conjugation Kit | NovaFluor™ Blue 760 |
K06T04L017 | Conjugation Kit | NovaFluor™ Red 660 |
K06T04L018 | Conjugation Kit | NovaFluor™ Red 685 |
K06T04L019 | Conjugation Kit | NovaFluor™ Red 700 |
K06T04L020 | Conjugation Kit | NovaFluro™ Red 710 |
K06T04L023 | Conjugation Kit | NovaFluor™ Red 725 |
K06T04L024 | Conjugation Kit | NovaFluor™ Red 755 |
K06T04L005 | Conjugation Kit | NovaFluor™ Yellow 570 |
K06T04L013 | Conjugation Kit | NovaFluor™ Yellow 590 |
K06T04L006 | Conjugation Kit | NovaFluor™ Yellow 610 |
K06T04L007 | Conjugation Kit | NovaFluor™ Yellow 660 |
K06T04L014 | Conjugation Kit | NovaFluor™ Yellow 690 |
K06T04L015 | Conjugation Kit | NovaFluor™ Yellow 700 |
K06T04L016 | Conjugation Kit | NovaFluor™ Yellow 730 |
K06T04L022 | Conjugation Kit | NovaFluor™ Yellow 755 |
K06T04L027 | Conjugation Kit | NovaFluor™ Blue 800 |
K06T04L028 | Conjugation Kit | NovaFluor™ Yellow 810 |
K06T04L029 | Conjugation Kit | NovaFluor™ Violet 800 |
K06T04L030 | Conjugation Kit | NovaFluor™ Violet 690 |
K06T04L031 | Conjugation Kit | NovaFluor™ Violet 745 |
K06T04L032 | Conjugation Kit | NovaFluor™ UV 765 |
K06T04L033 | Conjugation Kit | NovaFluor™ Blue 725 |
NovaFluor Conjugation Kit provides features including:
3-step conjugation method