1x1 image pixel for data collection
GeneJET Genomic DNA Purification Kit
GeneJET Genomic DNA Purification Kit
Thermo Scientific™
GeneJET Genomic DNA Purification Kit
Thermo Scientific GeneJET Genomic DNA Purification Kit is designed for rapid and efficient purification of high quality genomic DNA fromRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
K072150 Preps
K0722250 Preps
Catalog number K0721
Price (USD)
144.65
Online Exclusive
185.00
Save 40.35 (22%)
Each
Add to cart
Quantity:
50 Preps
Price (USD)
144.65
Online Exclusive
185.00
Save 40.35 (22%)
Each
Add to cart
Thermo Scientific GeneJET Genomic DNA Purification Kit is designed for rapid and efficient purification of high quality genomic DNA from various mammalian cell culture and tissue samples, whole blood, bacteria and yeast.
The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.
The standard procedure takes less than 20 minutes following cell lysis and yields purified DNA greater than 30 kb in size. Isolated DNA can be used directly in PCR, Southern blotting and enzymatic reactions.
Highlights
Universal—can be used for both cell and tissue samples from a wide range of sources
Efficient—high yields of high molecular weight genomic DNA
Fast—20 minute procedure following cell lysis
Convenient—spin columns are capped and assembled with collection tubes
Applications
Isolated genomic DNA is ideal for use in common molecular biology procedures, including:
• PCR and qPCR
• FastDigest and conventional restriction digestion
• Southern blotting
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeSpin Column
For Use With (Application)Real-Time Quantitative PCR (qPCR), Cloning, Southern Blotting, Sequencing, PCR
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Product LineGeneJET
Product TypeGenomic DNA Purification Kit
Quantity50 Preps
Sample TypeBacteria, Buccal Samples, Cells, Tissue, Yeast
TargetGenomic DNA
Test Time<20 min.
FormatSpin Column
Isolation TechnologySpin Column
Unit SizeEach
Frequently asked questions (FAQs)
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?
We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.
Can I use a vacuum manifold with GeneJET kits?
The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.
At what temperature should I perform centrifugation steps in the GeneJET kits?
We recommend performing centrifugation steps at room temperature when the temperature is not specified.
Are GeneJET spin columns or MagJET magnetic beads available separately?
No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection