Fast DNA End Repair Kit
Fast DNA End Repair Kit
Thermo Scientific™
Fast DNA End Repair Kit
Thermo Scientific Fast DNA End Repair Kit is used for blunting and phosphorylation of DNA ends in just 5 minutesRead more
Have Questions?
Catalog NumberQuantity
K077150 Reactions
Catalog number K0771
Price (USD)
241.65
Online Exclusive
265.00
Save 23.35 (9%)
Each
In stock
Add to cart
Quantity:
50 Reactions
Request bulk or custom format
Price (USD)
241.65
Online Exclusive
265.00
Save 23.35 (9%)
Each
Add to cart
Thermo Scientific Fast DNA End Repair Kit is used for blunting and phosphorylation of DNA ends in just 5 minutes for subsequent use in blunt-end ligation.
All components of the kit contain premixed reagents to reduce pipetting steps and provide convenience. The End Repair Enzyme Mix contains an optimized mixture of T4 DNA Polymerase and Klenow Fragment to achieve highly effective blunting of fragmented DNA, and T4 Polynucleotide Kinase (PNK) for efficient phosphorylation of DNA ends. The 10X End Repair Reaction Mix contains an optimized reaction buffer, ATP, and dNTPs.
Samples such as fragmented genomic DNA (restriction enzyme digested, nebulized or sonicated), restriction enzyme digested plasmid DNA, double stranded cDNA, and PCR products containing dA overhangs are all compatible with the kit.
Features
During the DNA end repair reaction, fragmented DNA is converted into blunt-end DNA containing a 5'-phosphate and 3'-hydroxyl groups. The 5'→3' polymerase activity of the End Repair Enzyme Mix fills-in 5' protruded DNA ends while 3'→5' exonuclease activity removes 3'-overhangs. T4 PNK adds 5'-phosphates to ends of unphosphorylated DNA fragments, such as PCR products. (see Table 1 and Figure 1 below).
Highlights
Efficient - blunting and phosphorylation of 0.5 to 5 μg DNA
Fast—reaction is completed in 5 minutes
Convenient—reaction components are premixed to reduce pipetting steps
Applications
• Blunting and phosphorylation of double-stranded DNA (nebulized DNA, sonicated DNA, restriction enzyme digested DNA, cDNA, PCR products)
Includes
• End Repair Enzyme Mix
• 10X End Repair Reaction Mix
Related Products
CloneJET PCR Cloning Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
EnzymeDNA Polymerase, T4 Polynucleotide Kinase
For Use With (Application)Cloning
Product TypeFast DNA End Repair Kit
Quantity50 Reactions
Sample TypeDNA
FormatKit
Unit SizeEach
Frequently asked questions (FAQs)
I recently ordered the Fast DNA End Repair Kit and noticed precipitates in the 10x reaction mix after thawing. Can I still use it?
It is possible for precipitates to form in the 10x reaction mix. The buffer contains DTT which can precipitate after freezing. We recommend resuspending it by vortexing the solution. This should only take a few minutes. We have observed no negative impact on the reaction if the DTT has precipitated and been resuspended again.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection