Informational:Informational:New optimized magnetic bead solution for DNA cleanup. Learn more here.
Informational:Informational:Need Ethanol or Isopropanol for your nucleic acid isolation? Visit our Molecular Biology Grade Solvents page.
Thermo Scientific™

GeneJET NGS Cleanup Kit

Catalog number: K0851
Have Questions?
Thermo Scientific™

GeneJET NGS Cleanup Kit

Catalog number: K0851
Have Questions?
Catalog Number
K0851
Unit Size
50 preps
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
250 preps
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K085150 preps
-
K0852250 preps
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific GeneJET NGS Cleanup Kit provides protocols optimized specially for fast DNA fragment cleanup or adapter removal procedures. The clean up protocol results in higher than 70% recovery of DNA fragments larger than 150 bp and more than 97% removal of adapters. Adapter removal protocol results in more than 70% recovery of DNA fragments larger than 200 bp and more than 99% removal of adapters. The entire procedure takes 5 minutes for DNA fragment library cleanup from enzymatic reaction mixtures and 7 minutes for adapter removal protocol. Spin columns technology in combination with unique binding buffer allows to get the reproducible yields of DNA fragments in the wide starting material range (5 ng - 1 μg of DNA fragments).

The GeneJET NGS Cleanup Kit is compatible with Illumina™, Life Technologies library preparation workflows.

Highlights
Fast—high quality DNA fragments obtained within 5 min
Robust—reliable cleanup and size selection from different reaction mixtures and buffers, such as end-conversion, ligation, adapter addition, PCR mixtures, fragmentation buffers
Efficient—DNA recovery up to 90%
NGS-grade purity—free of primers, adapters, nucleotides, enzymes and other reaction components inhibiting NGS workflow.

Applications
• Purified DNA fragments are ideal for use in:
• PCR and qPCR
• Next Generation Sequencing workflow

Includes
• Binding Buffer (Cat. No. R2811)
• Prewash Buffer (conc.) (Cat. No. R2821)
• Wash Buffer (conc.)(Cat. No. R2831)
• Elution Buffer (10 mM Tris-HCl, pH 8.5)
• DNA Purification Micro Column & Collection Tube
• Collection Tube

Related Products
GeneJET NGS Cleanup Kit
Binding Buffer for GeneJET NGS Cleanup Kit
Prewash Buffer for GeneJET NGS Cleanup Kit
Wash Buffer for GeneJET NGS Cleanup Kit

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Spin Column
Isolation Technology
Spin Column
Sample Type
DNA
For Use With (Application)
Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR), PCR
High-throughput Compatibility
Not High-throughput Compatible (Manual)
No. of Reactions
50 Preps
Product Line
GeneJET
Quantity
50 Preps
Target
DNA from Enzymatic Reactions (e.g. PCR)
Test Time
5 min.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text