Loading Solutions
Loading Solutions
Loading Solutions
This kit contains the solutions needed to conveniently load the FRET-based, beta-lactamase substrates, CCF2-AM and CCF4-AM, into cells.Read more
Have Questions?
Catalog NumberQuantity
K10261 Kit
Catalog number K1026
Price (USD)
530.00
Each
Estimated availability date 21-Aug-2025
Add to cart
Quantity:
1 Kit
Recurring order eligible. Learn more »
Price (USD)
530.00
Each
Add to cart
Ask our AI about this Product
This kit contains the solutions needed to conveniently load the FRET-based, beta-lactamase substrates, CCF2-AM and CCF4-AM, into cells.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineGeneBLAzer™
Quantity1 Kit
Shipping ConditionRoom Temperature
TypeLoading Solution
Unit SizeEach
Contents & Storage
Store at room temperature.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Frequently asked questions (FAQs)
Do you offer an alternative to GeneBLAzer In Vivo Detection Kit (Cat. No. 12578134)?
The following are two alternatives to the GeneBLAzer In Vivo Detection Kit:
- LiveBLAzer FRET-B/G Loading Kit with CCF2-AM (Cat. No. K1032, K1025)
- LiveBLAzer FRET-B/G Loading Kit with CCF4-AM (Cat. No. K1095, K1096, K1030)
Concerning CCF2-AM and CCF4-AM, these molecules differ by two carbons in the bridge that links the coumarin moiety to the lactam ring. Both are in the membrane-permeable, esterified forms, and can be used for assays in intact cells. CCF4-AM has better solubility properties than CCF2-AM and is thus best suited for screening applications. In addition, CCF4-AM has slightly better FRET and thus slightly lower background than CCF2-AM.
Note: We also offer the loading solutions as a separate kit, Loading Solutions (Cat. No. K1026), and CCF2-AM is also available as a standalone product (Cat. No. K1023).
Figures
Customers who viewed this item also viewed
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
2904390Certificate of AnalysisFeb 21, 2025K1026
3081354Certificate of AnalysisFeb 21, 2025K1026
2888021Certificate of AnalysisFeb 21, 2025K1026
2997056Certificate of AnalysisFeb 21, 2025K1026
3166987Certificate of AnalysisFeb 21, 2025K1026
5 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection