DreamTaq PCR Master Mixes (2X)
DreamTaq PCR Master Mixes (2X)
Thermo Scientific™

DreamTaq PCR Master Mixes (2X)

Thermo Scientific DreamTaq PCR Master Mix (2X) is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq buffer, MgCl2, and dNTPs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorQuantity
K1071Colorless4 x 1.25 mL
K1072Colorless20 x 1.25 mL
K1081Green4 x 1.25 mL
K1082Green20 x 1.25 mL
Catalog number K1071
Price (EUR)
123,65
Online Exclusive
141,00
Save 17,35 (12%)
Each
-
Add to cart
Color:
Colorless
Quantity:
4 x 1.25 mL
Request bulk or custom format
Price (EUR)
123,65
Online Exclusive
141,00
Save 17,35 (12%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific DreamTaq PCR Master Mix (2X), available in colorless and green formats, is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq buffer, MgCl2, and dNTPs. This pre-mixed formulation saves time and reduces contamination due to the fewer pipetting steps required for PCR setup.

The master mix retains all features of DreamTaq DNA Polymerase and is capable of robust amplification up to 6 kb from genomic DNA and up to 20 kb from viral DNA. It ensures higher sensitivity, longer PCR products, and higher yields compared to conventional Taq DNA polymerase. Extensive optimization of reaction conditions is not required.

DreamTaq Green PCR Master Mix (2X) additionally includes 2X DreamTaq Green Buffer with a density reagent and two dyes for monitoring electrophoresis progress.

Features

  • Robust amplification with minimal optimization
  • High yields of PCR products
  • Higher sensitivity compared to conventional Taq DNA polymerase
  • Amplification of targets up to 6 kb from genomic DNA and up to 20 kb from viral DNA
  • Incorporates modified nucleotides, but does not incorporate dUTP
  • Direct loading of PCR product on gel with green format

Applications

  • Routine PCR amplification of DNA fragments up to 6 kb
  • High throughput PCR
  • Genotyping
For Research Use Only. Not for use in diagnostic procedures.

In certain countries use of this product is covered by patents. Purchase of product in these countries includes non-transferable, limited license for using only this amount of product for the purchaser's own internal research.

This product is licensed under one or more U.S. Patents Nos. 5,500,363 and 5,352,778 or corresponding foreign patents. This product is licensed under one U.S. Patent No. 5,436,149 or corresponding foreign patents owned by Takara Bio, Inc.

Patent pending.

Specifications
ColorColorless
Concentration5 U/μL
Fidelity (vs. Taq)1X
Hot StartNo
Overhang3'-A
PolymeraseDreamTaq DNA Polymerase
Quantity4 x 1.25 mL
Reaction FormatSuperMix or Master Mix
Shipping ConditionDry Ice
For Use With (Application)Standard PCR
GC-Rich PCR PerformanceLow
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
• DreamTaq PCR Master Mix (2X), 4 x 1.25 mL
• Nuclease-free water, 4 x 1.25 mL

Store at -20°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link