Thermo Scientific™
GeneBLAzer™ Jurkat Control Kit
Catalog number: K1206
Have Questions?
Thermo Scientific™
GeneBLAzer™ Jurkat Control Kit
Catalog number: K1206
Have Questions?
Catalog Number
K1206
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K12061 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Th e GeneBLAzer™ Jurkat Control Kit contains constitutively expressing (CMV-bla) and wildtype Jurkat cells to provide positive and negative controls, respectively, for evaluating beta-lactamase activity (Figure 1).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell State
Dividing Cells
Detection Method
Fluorescent
Product Line
GeneBLAzer™
Cell Line
Jurkat
System Type
GeneBLAzer™
Druggable Target
Signaling Pathway
Contents & Storage
Th e GeneBLAzer™ Jurkat Control Kit contains two vials of cells, one ml per vial (∼ 5 x 106 cells/ml) and is shipped on dry ice. Store in liquid nitrogen upon receipt.