Search Thermo Fisher Scientific
The GeneBLAzer™ CHO-K1 Control Kit contains constitutively expressing (CMV-bla) CHO-K1 cells and wildtype CHO-K1 cells to provide positive and negative controls for evaluating beta-lactamase activity.