CloneJET PCR Cloning Kit
CloneJET PCR Cloning Kit
Thermo Scientific™

CloneJET PCR Cloning Kit

Thermo Scientific CloneJET PCR Cloning Kit, also available with DH10B cells (K123120), is an advanced positive selection system for high-efficiencyRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
K123120 Reactions
K123240 Reactions
Catalog number K1231
Price (USD)
178.65
Online Exclusive
208.00
Save 29.35 (14%)
Each
-
Add to cart
No. of Reactions:
20 Reactions
Request bulk or custom format
Price (USD)
178.65
Online Exclusive
208.00
Save 29.35 (14%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific CloneJET PCR Cloning Kit, also available with DH10B cells (K123120), is an advanced positive selection system for high-efficiency cloning of PCR products generated with any thermostable DNA polymerase. Any other blunt or sticky-end DNA fragment can be cloned. It is ideal for phosphorylated or non-phosphorylated DNA fragments. Ligation into the included positive selection vector takes only 5 minutes, yielding more than 99% recombinant clones. Blunt-ended PCR products generated with a proofreading enzyme are ligated directly into the cloning vector.

PCR products generated either with non-proofreading DNA polymerases or mixtures of DNA polymerases are blunted prior to ligation in 5 minutes with the thermostable DNA Blunting Enzyme provided with the kit. All common laboratory E. coli strains can be directly transformed with the ligation product.

Features

The CloneJET PCR Cloning Kit contains a novel, ready-to-use positive selection cloning vector pJET1.2/blunt. The vector contains a lethal restriction enzyme gene that is disrupted by ligation of a DNA insert into the cloning site. As a result, only bacterial cells with recombinant plasmids are able to form colonies. Recircularized pJET1.2/blunt vector molecules lacking an insert express a lethal restriction enzyme, which kills the host E. coli cell after transformation. This positive selection drastically accelerates the process of colony screening and eliminates additional costs required for blue/white selection.

For convenience in mapping and manipulation of the insert, the pJET1.2/blunt cloning vector multiple cloning site contains two BglII recognition sequences that flank the insertion site. In addition, the vector contains a T7 promoter for in vitro and in vivo transcription as well as sequencing of the insert.

Highlights

Fast—PCR cloning in only 5 minutes
Highest efficiency – > 99% of positive clones
No cloning background—positive selection vector
Versatile—ideal for blunt-end or sticky-end cloning
Economical—no expensive blue/white screening

Applications

• Cloning of blunt-end or 3'-dA tailed PCR products up to 10 kb
• Cloning of DNA fragments generated by restriction enzymes
• Sequencing of cloned DNA
in vitro and in vivo transcription of cloned inserts from the T7 promoter

Includes

• pJET1.2/blunt Cloning Vector
T4 DNA Ligase
• 2X Reaction Buffer
• DNA Blunting Enzyme
• pJET1.2 Forward Sequencing Primer (5'-CGACTCACTATAGGGAGAGCGGC-3')
• pJET1.2 Reverse Sequencing Primer (5'-AAGAACATCGATTTTCCATGGCAG-3')
• Control PCR Product
Water, nuclease-free
• Detailed Protocol

Prior to electroporation, always column-purify the ligation mixture using e.g. GeneJET PCR Purification Kit #K0701 or chloroform to extract it. Electroporation is inhibited by the presence of proteins and salts in the mixture.

Related Products
CloneJET PCR Cloning Kit
Fast DNA End Repair Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialAmpicillin (AmpR)
Cloning MethodRestriction Enzyme⁄ MCS, Blunt PCR
For Use With (Application)PCR Cloning
No. of Reactions20 Reactions
Product LineCloneJET
Product TypePCR Cloning Kit
Quantity20 Reactions
Sample TypePCR Products
VectorpJET1.2/blunt
FormatKit
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link