Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

CloneJET™ PCR Cloning Kit with DH10B Competent Cells

Catalog number: K123120
Have Questions?
Thermo Scientific™

CloneJET™ PCR Cloning Kit with DH10B Competent Cells

Catalog number: K123120
Have Questions?
Catalog Number
K123120
Unit Size
20 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
40 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K12312020 reactions
-
K12324040 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This package includes the CloneJET PCR Cloning Kit (20 or 40 reactions) and DH10B Competent Cells that are optimized for use with the kit and can be directly transformed with the ligation product.

CloneJET PCR Cloning Kit
The Thermo Scientific CloneJET PCR Cloning Kit is an advanced positive-selection system for high-efficiency cloning of PCR products generated with any thermostable DNA polymerase. Any blunt or sticky-end DNA fragment can be cloned. The kit works well with both phosphorylated and non-phosphorylated DNA fragments. Ligation into the included positive-selection vector takes only five minutes, yielding more than 99% recombinant clones. Blunt-ended PCR products generated with a proofreading enzyme are ligated directly into the cloning vector.

PCR products generated either with non-proofreading DNA polymerases or mixtures of DNA polymerases need to be blunted prior to ligation in a five minute reaction with the thermostable DNA Blunting Enzyme provided with the kit. DH10B Competent Cells are optimized for use with the CloneJET PCR Cloning Kit and can be directly transformed with the ligation product.

The CloneJET PCR Cloning Kit contains the novel, ready-to-use positive selection cloning vector pJET1.2/blunt. The vector contains a lethal restriction enzyme gene that is disrupted by ligation of a DNA insert into the cloning site. As a result, only bacterial cells with recombinant plasmids are able to form colonies. Re-circularized pJET1.2/blunt vector molecules lacking an insert express a lethal restriction enzyme, which kills the host E. coli cell after transformation. This positive selection drastically accelerates the process of colony screening and eliminates additional costs required for blue/white selection.

Fast—PCR cloning in only five minutes
Highest efficiency—>99% of positive clones
No cloning background—positive selection vector
Versatile—ideal for blunt-end or sticky-end cloning
Economical—no expensive blue/white screening

Applications include:
• Cloning of blunt-end or 3' dA-tailed PCR products up to 10 kb
• Cloning of DNA fragments generated by restriction enzymes
• Sequencing of cloned DNA
• in vitro and in vivo transcription of cloned inserts from the T7 promoter

Learn more about the CloneJET PCR Cloning Kit ›

DH10B Competent Cells
Thermo Scientific DH10B Competent Cells are high efficiency, chemically competent E.coli cells. The DH10B strain is suitable for cloning DNA containing methylcytosine and methyladenine, allowing both prokaryotic and eukaryotic genomic DNA to be cloned efficiently. These cells are ideal for the construction of cDNA libraries using plasmid-derived vectors. Genotype: F– mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 endA1 araD139 Δ (ara-leu)7697 galU galK λ– rpsL(StrR) nupG.

• High transformation efficiency: >1x109 cfu/μg pUC19 DNA
• Suitable for routine and high-throughput cloning applications
• Convenient two reactions per vial packaging
• S.O.C. Outgrowth medium included

Related products
CloneJET PCR Cloning Kit without competent cells
aLICator LIC Cloning and Expression System
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Bacterial or Yeast Strain
DH10B
Antibiotic Resistance Bacterial
Ampicillin (AmpR)
Cell Type
DH10B
Cloning Method
Blunt PCR, Restriction Enzyme ⁄ MCS
Format
Kit
Sample Type
PCR Products
Efficiency
≥1 x 10^9
For Use With (Application)
PCR Cloning
No. of Reactions
20 Reactions
Product Line
Thermo Scientific™
Product Type
PCR Cloning Kit
Quantity
20 Reactions
Shipping Condition
Dry Ice
Vector
pJET1.2/blunt

Contents & Storage

• 10 x DH10B Competent Cells for Subcloning (100 μL)
• pUC19 DNA (20 μL) (100 pg/μL)
• S.O.C. Medium (5 mL)
• pJET1.2/blunt Cloning Vector (24 μL) (50 ng/μL)
• 2X Reaction Buffer (300 μL)
• T4 DNA Ligase (24 μL) (5 U/μL)
• DNA Blunting Enzyme (24 μL)
• DNA Blunting Enzyme (24 μL)
• pJET1.2 Forward Sequencing Primer, 10 μM aqueous solution (50 μL)
• pJET1.2 Reverse Sequencing Primer, 10 μM aqueous solution (50 μL)
• Control PCR Product, 976 bp with 3’-dA overhangs (8 μL) (24 ng/μL)
• Water, nuclease-free, (1.25 mL)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text