Thermo Scientific™

MuSeek Library Preparation Kit, Illumina compatible

Catalog number: K1361
Have Questions?
Thermo Scientific™

MuSeek Library Preparation Kit, Illumina compatible

Catalog number: K1361
Have Questions?
Catalog Number
K1361
Unit Size
12 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K136112 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific™ MuSeek™ Library Preparation Kit, Illumina™ compatible is designed for generation of high‐quality genomic DNA libraries for sequencing with the Illumina™ MiSeq™ and HiSeq™ systems. The fast protocol utilizes the MuA transposase enzyme, which catalyzes simultaneous fragmentation of double-stranded target DNA and tagging the fragment ends with transposon DNA. In a subsequent PCR step the platform-specific adapters are added using a robust and accurate Thermo Scientific Phusion™ High-Fidelity DNA polymerase. Starting with only 50 ng or 100 ng of sample the protocol can be used to generate 100 bp - 1,000 bp insert libraries. The kit contains components sufficient for 12 fragmentation reactions and the subsequent adapter-addition polymerase chain reactions. Sequencing primers required for sequencing libraries prepared by this method are provided as well.

Highlights

Convenient—no physical shearing, end-repair and ligation steps
Fast—generates NGS fragment libraries in less than 90 minutes
Low sample input—DNA fragment libraries from 50 ng DNA input
Indexing compatible—MuSeek Index Set 1 enables preparation of up to 24 dual-indexed DNA libraries for sequencing in one run.

Applications

• DNA fragment library preparation for downstream use in next generation sequencing workflow.
• Library preparation from gDNA, ds DNA, bacterial DNA, DNA amplicons.

Includes
• MuSeek Enzyme Mix, IL
• MuSeek Fragmentation Reaction Buffer
• MuSeek Stop Solution
• Water, nuclease-free
• Control DNA
• MuSeek Adapter Addition Primer Mix, IL
• MuSeek Adapter Addition Reaction Buffer, IL
• Phusion™ Hot Start II High-Fidelity DNA Polymerase, IL
• MuSeek Sequencing Primer, Read 1
• MuSeek Sequencing Primer, Read 2
• MuSeek Sequencing Primer, Index Read

Notes
The kit is shipped on dry ice. Upon arrival store at -20°C, avoid repeated freeze-thaw cycles. The Stop Solution can be stored at room temperature.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Library Preparation Kit
Libraries
Fragment Library
For Use With (Equipment)
Illumina™ HiSeq 2500, Illumina ™ MiSeq
Sample Type
DNA
Sequencing Type
Genome & DNA Sequencing
Workflow Step
Library Generation
Product Line
MuSeek
For Use With (Application)
NGS library preparation
Quantity
12 Reactions

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text