GeneBLAzer™ RXR beta DA Assay Kit
Product Image
Thermo Scientific™

GeneBLAzer™ RXR beta DA Assay Kit

GeneBLAzer™RXR beta DA (Division Arrested) cells and RXR beta-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the humanRead more
Have Questions?
Catalog number K1405
Price (USD)/ 1 plate
1,492.00
-
Add to cart
Price (USD)/ 1 plate
1,492.00
Add to cart
GeneBLAzer™ RXR beta DA Assay Kit
Catalog numberK1405
Price (USD)/ 1 plate
1,492.00
-
Add to cart

GeneBLAzer™RXR beta DA (Division Arrested) cells and RXR beta-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Retinoid X Receptor beta (RXR beta) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line. GeneBLAzer™UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-RXR beta (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both RXR beta DA cells and RXR beta-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of 9-cis retinoic acid (9-cis-RA). In addition, RXR beta-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, and stimulation time.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Alternate AgonistsProgesterone
AntagonistsRU 486
Assay EntryCell-based beta lactamase reporter gene
Cell StateDivision-Arrested Cells
Detection MethodFluorescence
Gene SymbolRXRβ
Primary AgonistR5020
Product LineGeneBLAzer™
Quantity1 Plate
Reporter GeneBLA (Beta-Lactamase)
Target EntryRXR beta, NR2B2, RXRB
Cell Line293T (HEK)
System TypeGeneBLAzer™
Druggable TargetNuclear Receptors
Unit Size1 plate
Contents & Storage
Contains sufficient division-arrested cells and substrate to assay 1 x 384-well plate:
• RXR beta HEK 293T DA cells (K1405A)
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen)