Thermo Scientific Rapid DNA Ligation Kit enables fast sticky-end or blunt-end DNA ligation in only 5 minutes at room temperature.The kit contains T4 DNA ligase and a specially-formulated 5X rapid ligation buffer optimized for fast and efficient DNA ligation. Fast ligation efficiency is equal to that obtained with T4 DNA ligase in a standard 1 hour ligation. The ligation reaction mixture can be used directly for bacterial transformation with the TransformAid Bacterial Transformation Kit or with other conventional transformation procedures.
Highlights
• Fast—ligation of either sticky- or blunt-end DNA in only 5 minutes • Convenient—reaction mixture can be used directly for bacterial transformation
Applications
• Routine cloning experiments • Blunt-end cloning • Library construction • TA cloning
Prior to electroporation, it is necessary to inactivate T4 DNA ligase by chloroform extraction or spin column purification, e.g. with Thermo Scientific GeneJET PCR Purification Kit (K0701).