GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 Cells
Product Image
Thermo Scientific™

GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 Cells

The GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 cells contain the human G-Protein Receptor 119 (GPR119), (Accession No. NP_848566) stably integrated into theRead more
Have Questions?
Catalog number K1486
Price (USD)/ 2 x 106 cells
10,580.00
-
Add to cart
Price (USD)/ 2 x 106 cells
10,580.00
Add to cart
GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 Cells
Catalog numberK1486
Price (USD)/ 2 x 106 cells
10,580.00
-
Add to cart

The GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 cells contain the human G-Protein Receptor 119 (GPR119), (Accession No. NP_848566) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. No. K1129) contain a beta-lactamase (bla) reporter gene under control of the cAMP response element.

The T-Rex™ GPR119-CRE-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of AR231453(1). In addition, T-Rex™ GPR119-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_178471.1
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Detection MethodFluorescence
Gene SymbolGPR119
Population StatusSingle Cell Clone Origin
Product LineGeneBLAzer™
Quantity2 x 106 Cells
Reporter GeneBLA (Beta-Lactamase)
Shipping ConditionRoom Temperature
Target EntryGPR119
Cell LineCHO-K1
System TypeGeneBLAzer™
Druggable TargetGPCR
Unit Size2 x 106 cells
Contents & Storage
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 cells
• GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.