RevertAid First Strand cDNA Synthesis Kit
RevertAid First Strand cDNA Synthesis Kit
Thermo Scientific™

RevertAid First Strand cDNA Synthesis Kit

Thermo Scientific RevertAid First Strand cDNA Synthesis Kit is a complete system for efficient synthesis of first strand cDNA fromRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
K162120 Reactions
K1622100 Reactions
Catalog number K1621
Price (USD)
108.65
Online Exclusive
137.00
Save 28.35 (21%)
Each
-
Add to cart
No. of Reactions:
20 Reactions
Request bulk or custom format
Price (USD)
108.65
Online Exclusive
137.00
Save 28.35 (21%)
Each
Add to cart

Thermo Scientific RevertAid First Strand cDNA Synthesis Kit is a complete system for efficient synthesis of first strand cDNA from RNA templates. The kit uses RevertAid Reverse Transcriptase (RT), a recombinant M-MuLV RT which maintains activity at 42–50°C and is suitable for synthesis of cDNA up to 13 kb. RiboLock RNase Inhibitor, supplied with the kit, effectively protects RNA templates from degradation at temperatures up to 55°C.

The RevertAid First Strand cDNA Synthesis Kit is supplied with both oligo(dT)18 and random hexamer primers. The oligo(dT)18 primer anneals selectively to the poly(A) tail of mRNA. Random hexamer primers do not require the presence of the poly(A) tail; they can therefore be used for transcription of the 5'-end regions of mRNA or cDNA synthesis of RNA species lacking a poly(A) tail (e.g., microRNAs). Gene-specific primers may also be used with the kit.

Radioactively- and non-radioactively-labeled nucleotides can be incorporated into first strand cDNA for use as a probe in hybridization experiments, including microarrays.

Features of the RevertAid First Strand cDNA Synthesis Kit include:
• Full-length first strand cDNA up to 13 kb
• Optimum reaction temperature 42°C
• Complete kit with all components necessary for the RT reaction already included

Applications
• First strand cDNA synthesis for RT-PCR and RT-qPCR
• Construction of full length cDNA libraries
• Antisense RNA synthesis

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeFirst-Strand cDNA
FormatKit
No. of Reactions20 Reactions
Optimal Reaction Temperature42°C
Quantity20 reactions
Reaction FormatSeparate components
Reagent TypeReverse Transcription
Reverse TranscriptaseRevertAid
Ribonuclease H ActivityYes
Shipping ConditionDry Ice
Size (Final Product)Up to 13 kb
Starting MaterialRNA
TechniqueReverse Transcription
For Use With (Application)RT-PCR
Reaction Speed60 min.
Unit SizeEach
Contents & Storage

• RevertAid Reverse Transcriptase
• RiboLock RNase Inhibitor
• 5X Reaction Buffer
• dNTP Mix
• Oligo(dT)18 Primer
• Random Hexamer Primer
• Control GAPDH RNA
• Forward GAPDH Primer (10 μM)
• Reverse GAPDH Primer (10 μM)
• Nuclease-free water

Store at –20°C.

Frequently asked questions (FAQs)

When should I choose regular RevertAid RT or Maxima RT vs. RevertAid H-minus RT or Maxima H-minus RT?

It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. It is also recommended to use RNase H-minus RTs for template-independent addition of C nucleotides. In contrast, reverse transcriptases with intrinsic RNase H activity are often favored in qPCR applications.

Do Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H-minus RT, Maxima RT, and Maxima H-minus RT) possess terminal deoxynucleotidyl (TdT) activity?

All Thermo Scientific reverse transcriptases possess intrinsic TdT activity although at varying degrees depending upon the reaction conditions. For addition of template-independent C nucleotides (as for SMART and RACE experiments), this specific TdT activity can be induced by Mn2+. We would recommend Maxima H- or RevertAid H- minus RTs for this purpose.

What steps should I take while performing first strand cDNA synthesis using low purity template (e.g., inhibitors in RNA sample)?

Trace amounts of reagents used in RNA purification protocols may remain in solution and inhibit first-strand synthesis, e.g., SDS, EDTA, guanidine salts, phosphate, pyrophosphate, polyamines, spermidine. To remove trace contaminants, we recommend re-precipitating the RNA with ethanol and washing the pellet with 75% ethanol, or re-purifying the RNA.

For reverse transcription, how important is the quality of RNA template?

RNA purity and integrity are essential for synthesis and quantification of cDNA. Always assess the integrity of RNA prior to cDNA synthesis. Use freshly prepared RNA. Multiple freeze/thaw cycles of the RNA sample and synthesized cDNA is not recommended. Avoid RNase contamination and discard low quality RNA.

When should I choose regular RevertAid RT or Maxima RT vs. RevertAid H Minus RT or Maxima H Minus RT?

It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. It is also recommended to use RNase H Minus RTs for template-independent addition of C nucleotides. In contrast, reverse transcriptases with intrinsic RNase H activity are often favored in qPCR applications.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link